Learn More
MSC Tanis Disc Brush Drive Arbor, 3-5 in Brush, 1 ea
Catalog No. 502866668
Encompass
The disc brush drive arbor is used to securely mount disc brushes for use in any type of milling or CNC machine. It facilitates coolant flow through the shaft and bolt.
  • Drive arbor connection type
  • Compatible with Tanis disc brushes
  • Features flow-through spindle for coolant
  • Includes bolt and washer
  • Shaft diameter of ¾ in
  • Designed for 3 in, 4 in, and 5 in brush diameters
  • Equipped with 2 locator pins
Catalog No. 50-286-6668 Supplier MSC Supplier No. 31890692
The disc brush drive arbor is used to securely mount disc brushes for use in any type of milling or CNC machine. It facilitates coolant flow through the shaft and bolt.
  • Drive arbor connection type
  • Compatible with Tanis disc brushes
  • Features flow-through spindle for coolant
  • Includes bolt and washer
  • Shaft diameter of ¾ in
  • Designed for 3 in, 4 in, and 5 in brush diameters
  • Equipped with 2 locator pins