Learn More
MSC Tanis Mini Disc Brush Drive Arbor, 1/2 in., 4500 RPM
Catalog No. 502867944
Encompass
The Mini Disc Brush Drive Arbor is engineered for securely mounting mini disc brushes onto milling or CNC machines. It facilitates efficient operation by ensuring a stable connection.
  • Securely mounts mini discs
  • Shaft diameter of 1/2 in.
  • Compatible with 2 in. and 3 in. brush diameters
  • Maximum operating speed of 4,500 RPM
  • Features flow-through coolant capability
  • Includes set screws for brush attachment
Catalog No. 50-286-7944 Supplier MSC Supplier No. 31890700
The Mini Disc Brush Drive Arbor is engineered for securely mounting mini disc brushes onto milling or CNC machines. It facilitates efficient operation by ensuring a stable connection.
  • Securely mounts mini discs
  • Shaft diameter of 1/2 in.
  • Compatible with 2 in. and 3 in. brush diameters
  • Maximum operating speed of 4,500 RPM
  • Features flow-through coolant capability
  • Includes set screws for brush attachment