Learn More
Key Scientific Products DISC FOR ONPG TESTING PK/50
SDP
Catalog No. NC0453501
Encompass_Preferred
Differentiate slow lactose fermenters and non-fermenters. Demonstration of beta-galactosidase accomplished by hydrolization of substrate that liberates ortho-nitrophenol for a characteristic yellow color. Includes 50 tests per pack.
Catalog No. NC0453501 Supplier Key Scientific Products Supplier No. K490
Differentiate slow lactose fermenters and non-fermenters. Demonstration of beta-galactosidase accomplished by hydrolization of substrate that liberates ortho-nitrophenol for a characteristic yellow color. Includes 50 tests per pack.