Learn More
Electron Microscopy Sciences Disc Gripper for AFM, 15 mm
SDP
Catalog No. 501930056
Encompass_Preferred
Use these unique grippers to easily pick up AFM/STM discs from a flat surface. Features a protective rubber-coated tip. Available in 4 sizes.
Size: 15 mm
Catalog No. 50-193-0056 Supplier Electron Microscopy Sciences Supplier No. 7500915
Use these unique grippers to easily pick up AFM/STM discs from a flat surface. Features a protective rubber-coated tip. Available in 4 sizes.
Size: 15 mm