Learn More
Electron Microscopy Sciences Disc Gripper for AFM, 20 mm
SDP
Catalog No. 501930057
Encompass_Preferred
Use these unique grippers to easily pick up AFM/STM discs from a flat surface. Features a protective rubber-coated tip. Available in 4 sizes.
Size: 20 mm
Catalog No. 50-193-0057 Supplier Electron Microscopy Sciences Supplier No. 7500920
Use these unique grippers to easily pick up AFM/STM discs from a flat surface. Features a protective rubber-coated tip. Available in 4 sizes.
Size: 20 mm