Learn More
Electron Microscopy Sciences Disc Holder
SDP
Catalog No. 50996838
Encompass_Preferred
Use with all diamond polishing discs and final discs. The surface is designed to hold disc securely and smoothly, with 1/8" (3.18mm) mandrel, maximum 12,000 RPM.
Catalog No. 50-996-838 Supplier Electron Microscopy Sciences Supplier No. 6214601
Use with all diamond polishing discs and final discs. The surface is designed to hold disc securely and smoothly, with 1/8" (3.18mm) mandrel, maximum 12,000 RPM.