Learn More
Perkin Elmer US LLC Infrared Window for Microscopic FTIR Analysis - KBr, 2 mm
Catalog No. 509045893
Encompass_Preferred
Infrared windows allow for support of microsamples for infrared analysis with mIcroscopes, such as PerkinElmer's Spotlight Systems. These windows are available in the following infrared crystals: BaF2, KBr, NaCl, and ZnSe.
Catalog No. 50-904-5893 Supplier Perkin Elmer US LLC Supplier No. N9302615
Infrared windows allow for support of microsamples for infrared analysis with mIcroscopes, such as PerkinElmer's Spotlight Systems. These windows are available in the following infrared crystals: BaF2, KBr, NaCl, and ZnSe.