Learn More
MSC Clarke Rider Autoscrubber, 28 inch Cleaning Path, Disc, Battery Powered
Catalog No. 502838873
Encompass
The Clarke Focus II Rider Autoscrubber is a productive and maneuverable floor cleaning machine designed for efficient cleaning of large areas. It features a 28 inch cleaning path and disc scrub head, offering cost-effective, sustainable performance and incorporating BOOST technology.
  • Tight turning radius.
  • Increased down pressure (up to 220 pounds) for one-pass cleaning.
  • Quiet operation.
  • Large 31-gallon tanks for extended scrubbing.
  • BOOST technology reduces chemical and water usage, extends pad life, and removes finishes without chemicals.
  • Low-profile SafetyGlide deck eliminates water marks.
  • Universal one-touch controls.
  • Safe-T-Steering for safety.
  • Durable squeegee design ensures excellent water pickup.
Catalog No. 50-283-8873 Supplier MSC Supplier No. 35706464
Add to Cart
Add to Cart
Request a Quote
The Clarke Focus II Rider Autoscrubber is a productive and maneuverable floor cleaning machine designed for efficient cleaning of large areas. It features a 28 inch cleaning path and disc scrub head, offering cost-effective, sustainable performance and incorporating BOOST technology.
  • Tight turning radius.
  • Increased down pressure (up to 220 pounds) for one-pass cleaning.
  • Quiet operation.
  • Large 31-gallon tanks for extended scrubbing.
  • BOOST technology reduces chemical and water usage, extends pad life, and removes finishes without chemicals.
  • Low-profile SafetyGlide deck eliminates water marks.
  • Universal one-touch controls.
  • Safe-T-Steering for safety.
  • Durable squeegee design ensures excellent water pickup.