Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MSC Clarke Focus II Disc Ride-On Battery Floor Scrubber, 34 inch Cleaning Path, 31 gal Solution Tank, 31 gal Recovery Tank

Catalog No. 502836651
Encompass

The Clarke Focus II Disc Ride-On Battery Floor Scrubber is an automatic scrubber designed for efficient floor maintenance. It offers a flexible and easy-to-use solution for both daily cleaning and demanding restoration tasks, featuring large tank capacities, maintenance-free batteries, and an onboard charger. This scrubber ensures sustainable performance and consistent results.

  • Rider automatic scrubber with a 34 inch disc cleaning path
  • Maintenance-free (AGM) batteries (312 Ah) and onboard charger
  • 31 gal solution and recovery tank capacities
  • 24 V power source
  • Low sound pressure level (68.6 dB A)
  • 46.1 in water lift
  • ETL/CSA agency approvals
  • Cleaning capacity of 46,675 ft²/hr with 3 hr battery run time
  • 3.12 mph speed with traction drive system

Catalog No. 50-283-6651 Supplier MSC Supplier No. 35706530
May include imposed supplier surcharges.
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

The Clarke Focus II Disc Ride-On Battery Floor Scrubber is an automatic scrubber designed for efficient floor maintenance. It offers a flexible and easy-to-use solution for both daily cleaning and demanding restoration tasks, featuring large tank capacities, maintenance-free batteries, and an onboard charger. This scrubber ensures sustainable performance and consistent results.

  • Rider automatic scrubber with a 34 inch disc cleaning path
  • Maintenance-free (AGM) batteries (312 Ah) and onboard charger
  • 31 gal solution and recovery tank capacities
  • 24 V power source
  • Low sound pressure level (68.6 dB A)
  • 46.1 in water lift
  • ETL/CSA agency approvals
  • Cleaning capacity of 46,675 ft²/hr with 3 hr battery run time
  • 3.12 mph speed with traction drive system

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.