Learn More
MSC Clarke Ride-On Floor Scrubber, 34 in Disc, 31 gal Tank
Catalog No. 502839294
Encompass
This mid-size rider scrubber delivers efficient and sustainable floor cleaning for daily maintenance and restoration tasks. It features large tank capacities, AGM batteries, optional chemical mixing, onboard charging, one-touch scrubbing, and adjustable squeegees.
  • 34 in disc cleaning path.
  • Four 312 Ah AGM batteries with onboard charger.
  • Optional BOOST technology for chemical-free finish removal.
  • One-touch scrubbing controls and adjustable squeegees.
  • Tight turning radius and increased down pressure.
  • Quiet operation.
  • 31 gal solution and recovery tanks for extended use.
  • Open machine design and tool-free adjustable seat.
  • Heavy-duty steel side skirts and non-marking polyurethane tires.
  • CSA, ETL Canada and US approved.
Catalog No. 50-283-9294 Supplier MSC Supplier No. 35706522
Add to Cart
Add to Cart
Request a Quote
This mid-size rider scrubber delivers efficient and sustainable floor cleaning for daily maintenance and restoration tasks. It features large tank capacities, AGM batteries, optional chemical mixing, onboard charging, one-touch scrubbing, and adjustable squeegees.
  • 34 in disc cleaning path.
  • Four 312 Ah AGM batteries with onboard charger.
  • Optional BOOST technology for chemical-free finish removal.
  • One-touch scrubbing controls and adjustable squeegees.
  • Tight turning radius and increased down pressure.
  • Quiet operation.
  • 31 gal solution and recovery tanks for extended use.
  • Open machine design and tool-free adjustable seat.
  • Heavy-duty steel side skirts and non-marking polyurethane tires.
  • CSA, ETL Canada and US approved.