Learn More
MSC Clarke Ride-On Orbital Floor Scrubber, 28 in, 31 gal, Chemical Mixing
Catalog No. 502838729
Encompass
This ride-on orbital floor scrubber provides powerful and efficient cleaning for commercial and industrial settings. Featuring a 28 inch cleaning path and 31 gallon solution and recovery tanks, it is ideal for large areas. An integrated chemical mixing system ensures precise solution proportioning, optimizing chemical use and enhancing cleaning effectiveness.
  • 28 inch cleaning path with coverage up to 38,438 sq. ft. per hour.
  • Adjustable down pressure (90, 150, 220 lb.) for varied cleaning.
  • Integrated chemical mixing system delivers precise solution.
  • Onboard charger and battery power for cordless flexibility.
  • 260 RPM brush speed for deep cleaning on diverse floors.
  • Easy maintenance with large recovery tank opening and debris catch cage.
  • Intuitive one-touch control panel simplifies operation.
  • ETL and CSA certified for safety standards.
Catalog No. 50-283-8729 Supplier MSC Supplier No. 35706589
Add to Cart
Add to Cart
Request a Quote
This ride-on orbital floor scrubber provides powerful and efficient cleaning for commercial and industrial settings. Featuring a 28 inch cleaning path and 31 gallon solution and recovery tanks, it is ideal for large areas. An integrated chemical mixing system ensures precise solution proportioning, optimizing chemical use and enhancing cleaning effectiveness.
  • 28 inch cleaning path with coverage up to 38,438 sq. ft. per hour.
  • Adjustable down pressure (90, 150, 220 lb.) for varied cleaning.
  • Integrated chemical mixing system delivers precise solution.
  • Onboard charger and battery power for cordless flexibility.
  • 260 RPM brush speed for deep cleaning on diverse floors.
  • Easy maintenance with large recovery tank opening and debris catch cage.
  • Intuitive one-touch control panel simplifies operation.
  • ETL and CSA certified for safety standards.