Learn More
ECM Bioscience DISC1 Phospho-Regulation Antibody Sampler Kit, ECM Biosciences
Supplier:  ECM Bioscience DK6690
Encompass_Preferred
The DISC1 phospho-regulation antibody kit can be used to detect phosphorylation of Ser-710 relative to total DISC1 expression. The kit includes two rabbit polyclonal antibodies to detect total expression of DISC1 and secondary reagents for detection of rabbit polyclonal antibody in western blot or ELISA. In addition, the kit includes peptides for blocking experiments or use as ELISA positive controls. Applications: WB, E; Species Reactivity: Rt, Ms
Catalog No. 50-206-8688