Learn More
AdipoGen DiSC35
SDP
Catalog No. 502561222
Encompass_Preferred
Chemical. CAS 53213-94-8. Formula C25H27IN2S2. MW 546.53. Synthetic. 3,3-Dipropylthiadicarbocyanine iodide is applied for transmembrane potential measurements.
Catalog No. 50-256-1222 Supplier AdipoGen Supplier No. CDXD0130G001
Chemical. CAS 53213-94-8. Formula C25H27IN2S2. MW 546.53. Synthetic. 3,3-Dipropylthiadicarbocyanine iodide is applied for transmembrane potential measurements.