Learn More
BURKLE INC Discharge hose with stopcock, PumpMaster petrochem.
Catalog No. 5000025845
Encompass_Preferred
This special discharge hose with shut-off valve was developed as an accessory for the Bürkle PumpMaster in blue — the drum pump designed for petrochemical liquids. Measuring 1.5 meters in length and featuring an integrated shut-off valve at the end, it allows for convenient and safe dispensing of mineral oils, diesel, or other non-flammable petrochemical liquids directly from the drum. Its robust construction ensures the hose remains shape-stable even with frequent use and guarantees secure transfer without leaks.
Catalog No. 50-000-25845 Supplier BURKLE INC Supplier No. 52023001
This special discharge hose with shut-off valve was developed as an accessory for the Bürkle PumpMaster in blue — the drum pump designed for petrochemical liquids. Measuring 1.5 meters in length and featuring an integrated shut-off valve at the end, it allows for convenient and safe dispensing of mineral oils, diesel, or other non-flammable petrochemical liquids directly from the drum. Its robust construction ensures the hose remains shape-stable even with frequent use and guarantees secure transfer without leaks.