Learn More
BURKLE INC Discharge hose with stopcock, PumpMaster acids
Catalog No. 5000025938
Encompass_Preferred
This discharge hose with shut-off valve is designed as an accessory for the green PumpMaster pump (FKM seals), which is developed for acids and aggressive chemicals.
With its integrated shut-off valve, it enables the safe dispensing of corrosive liquids such as acids, alkalis, or chemicals from canisters and drums.
The shut-off valve at the end allows the flow to be quickly stopped at any time, significantly enhancing safety when handling hazardous liquids.
Catalog No. 50-000-25938 Supplier BURKLE INC Supplier No. 52021001
This discharge hose with shut-off valve is designed as an accessory for the green PumpMaster pump (FKM seals), which is developed for acids and aggressive chemicals.
With its integrated shut-off valve, it enables the safe dispensing of corrosive liquids such as acids, alkalis, or chemicals from canisters and drums.
The shut-off valve at the end allows the flow to be quickly stopped at any time, significantly enhancing safety when handling hazardous liquids.