Learn More
BURKLE INC Discharge hose with stopcock, PumpMaster liquids
Catalog No. 5000025612
Encompass_Preferred
This drain hose with shut-off valve is optimized for use with the red PumpMaster pump (featuring EPDM seals) for transferring non-aggressive, water-based liquids.
With a hose length of 1.5 meters and an easy-to-use outlet valve, it allows for convenient dispensing of water, diluted solutions, or other mildly aggressive media from drums and containers.
The shut-off valve enables complete flow control, allowing the liquid flow to be stopped entirely.
Catalog No. 50-000-25612 Supplier BURKLE INC Supplier No. 52022001
This drain hose with shut-off valve is optimized for use with the red PumpMaster pump (featuring EPDM seals) for transferring non-aggressive, water-based liquids.
With a hose length of 1.5 meters and an easy-to-use outlet valve, it allows for convenient dispensing of water, diluted solutions, or other mildly aggressive media from drums and containers.
The shut-off valve enables complete flow control, allowing the liquid flow to be stopped entirely.