Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Discoidin domain-containing receptor 2, rat | >95.0% | 5 UG
SDP

Catalog No. 5000291049
Encompass_Preferred

DDR2 is a recombinant rat-derived discoidin domain receptor 2 protein expressed in HEK293 cells with a C-terminal His tag. It is a receptor tyrosine kinase involved in tissue remodeling and extracellular matrix signaling, supplied for research applications such as biochemical assays, signaling studies, and antibody validation.

  • Expressed in HEK293 cells.
  • Derived from rat sequence.
  • C-terminal His tag for detection and purification.
  • Purity greater than 95% as determined by reducing SDS-PAGE.
  • Available in small research quantities suitable for assay development.
  • Suitable for biochemical assays, signaling studies, and antibody validation.

Catalog No. 50-002-91049 Supplier Medchemexpress LLC Supplier No. HYP752975UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

DDR2 is a recombinant rat-derived discoidin domain receptor 2 protein expressed in HEK293 cells with a C-terminal His tag. It is a receptor tyrosine kinase involved in tissue remodeling and extracellular matrix signaling, supplied for research applications such as biochemical assays, signaling studies, and antibody validation.

  • Expressed in HEK293 cells.
  • Derived from rat sequence.
  • C-terminal His tag for detection and purification.
  • Purity greater than 95% as determined by reducing SDS-PAGE.
  • Available in small research quantities suitable for assay development.
  • Suitable for biochemical assays, signaling studies, and antibody validation.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.