Learn More
MilliporeSigma™ Supelco™ Discovery™ DSC-18 SPE 96-Well Plate, Bed Wt. 25 mg/well
Catalog No. 111008915 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Bed Weight:
25 mg/Well
Catalog No. Bed Weight
11-100-8915 25 mg/Well
1 options
Catalog No. 11-100-8915 Supplier MilliporeSigma™ Supelco™ Supplier No. 575601U
Add to Cart
Add to Cart
Solid phase extraction (SPE) is a technique commonly used in laboratories to isolate analytes of interest from complex matrices.
Retention Mechanism: Reversed-phase. Sample Matrix Compatibility: Aqueous solutions (biological fluids, water). Polymerically bonded, octadecyl (18 %C), endcapped. Higher 18 %C loading for increased binding capacities and higher recoveries. The least selective phase: retains most organic analytes from aqueous matrices. Can also be used for desalting aqueous matrices. Beneficial for extracting structurally diverse analytes from the same sample.
TRUSTED_SUSTAINABILITY
Specifications
Column Phase DSC-18
Column Type SPE
Bed Weight 25 mg/Well
For Use With (Application) Food and Beverages
Product Line Discovery
Type 96-well Plate
Volume (Metric) 2 mL
Endcapped Yes
Material Polypropylene
Pore Size 70 Å pore size
Quantity 1/Pk.
Surface Area (Metric) 480 m 2 /g
Show More Show Less
Discovery is a registered trademark of Sigma-Aldrich Co. LLC