Learn More
MilliporeSigma™ Supelco™ Discovery™ DSC-18 SPE Bulk Packing
Catalog No. 111003703 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
100 g
Catalog No. Quantity
11-100-3703 100 g
1 options
Catalog No. 11-100-3703 Supplier MilliporeSigma™ Supelco™ Supplier No. 52600U
Add to Cart
Add to Cart
Endcapped
Retention Mechanism: Reversed-phase. Sample Matrix Compatibility: Aqueous solutions (biological fluids, water). Polymerically bonded, octadecyl (18 %C), endcapped. Higher 18 %C loading for increased binding capacities and higher recoveries. The least selective phase: retains most organic analytes from aqueous matrices. Can also be used for desalting aqueous matrices. Beneficial for extracting structurally diverse analytes from the same sample.
TRUSTED_SUSTAINABILITY
Specifications
Pore Size 70 Å
Quantity 100 g
Surface Area (Metric) 480 m2/g
Product Line Discovery
Discovery is a registered trademark of Sigma-Aldrich Co. LLC