Learn More
TA Instruments Discovery DSC 25 Auto, General Purpose DSC with Autosampler, Fusion Cell, Max Temp 725°C, Min Temp -180°C with LN Cooling, Touchscreen, Modulated DSC (MDSC), TRIOS Software, 5-Year Warranty on DSC Cells
Catalog No. 5000580755 Shop All Waters TA Instruments Products
Encompass_Preferred
Waters TA Instruments Discovery DSC 25 Auto is a powerful, general-purpose differential scanning calorimeter designed for high-performance thermal analysis. Featuring the advanced Fusion Cell, it delivers exceptional baseline flatness, sensitivity, resolution, and reproducibility—without the need for data manipulation. The system supports a wide temperature range, from -180°C with liquid nitrogen cooling to 725°C, and includes a reliable linear autosampler for flexible, unattended operation. Standard features include a modern touchscreen interface with One-Touch-Away functionality, Modulated DSC (MDSC) for separating complex thermal events, and TRIOS software for intuitive experiment control, data analysis, and automated calibration. TRIOS is compatible with Windows 7 and newer. The DSC 25 Auto includes a five-year warranty on the DSC cell. Installation requires a DSL Router with 4-Port Switch (P/N: 251471.002) and Universal AC Adapter Kit (P/N: 203599.901) for international use.
Catalog No. 50-005-80755 Supplier TA Instruments Supplier No. 973700.904
Waters TA Instruments Discovery DSC 25 Auto is a powerful, general-purpose differential scanning calorimeter designed for high-performance thermal analysis. Featuring the advanced Fusion Cell, it delivers exceptional baseline flatness, sensitivity, resolution, and reproducibility—without the need for data manipulation. The system supports a wide temperature range, from -180°C with liquid nitrogen cooling to 725°C, and includes a reliable linear autosampler for flexible, unattended operation. Standard features include a modern touchscreen interface with One-Touch-Away functionality, Modulated DSC (MDSC) for separating complex thermal events, and TRIOS software for intuitive experiment control, data analysis, and automated calibration. TRIOS is compatible with Windows 7 and newer. The DSC 25 Auto includes a five-year warranty on the DSC cell. Installation requires a DSL Router with 4-Port Switch (P/N: 251471.002) and Universal AC Adapter Kit (P/N: 203599.901) for international use.