Learn More
TA Instruments Discovery DSC 25, Cost-Effective, General Purpose DSC, Fusion Cell Technology, Max Temp 725°C, Min Temp -180°C with Liquid Nitrogen Cooling, Touchscreen, Modulated DSC (MDSC), TRIOS Software, 5-Year Warranty on DSC Cells
Catalog No. 5000580619 Shop All Waters TA Instruments Products
Encompass_Preferred
Waters TA Instruments Discovery DSC 25 is a cost-effective, general-purpose differential scanning calorimeter designed for high-performance thermal analysis. At its core is the advanced Fusion Cell, delivering best-in-class baseline flatness, sensitivity, resolution, and reproducibility—without the need for data manipulation. The DSC 25 supports a wide temperature range, from -180°C with liquid nitrogen cooling to 725°C, depending on the cooling accessory. Standard features include a modern touchscreen interface with One-Touch-Away functionality, Modulated DSC (MDSC) for separating complex thermal events, and TRIOS software for intuitive experiment control, data analysis, and calibration. TRIOS is compatible with Windows 7 and newer. The Discovery DSC 25 includes a five-year warranty on the DSC cell. Installation requires a DSL Router with 4-Port Switch (P/N: 251471.002) and Universal AC Adapter Kit (P/N: 203599.901) for international use.
Catalog No. 50-005-80619 Supplier TA Instruments Supplier No. 973700.903
Waters TA Instruments Discovery DSC 25 is a cost-effective, general-purpose differential scanning calorimeter designed for high-performance thermal analysis. At its core is the advanced Fusion Cell, delivering best-in-class baseline flatness, sensitivity, resolution, and reproducibility—without the need for data manipulation. The DSC 25 supports a wide temperature range, from -180°C with liquid nitrogen cooling to 725°C, depending on the cooling accessory. Standard features include a modern touchscreen interface with One-Touch-Away functionality, Modulated DSC (MDSC) for separating complex thermal events, and TRIOS software for intuitive experiment control, data analysis, and calibration. TRIOS is compatible with Windows 7 and newer. The Discovery DSC 25 includes a five-year warranty on the DSC cell. Installation requires a DSL Router with 4-Port Switch (P/N: 251471.002) and Universal AC Adapter Kit (P/N: 203599.901) for international use.