Learn More
TA Instruments Discovery DSC 25P Accessory Kit, Includes Silver Cell Lids, Indium Calibration, Aluminum Pans and Lids, Sapphire Calibration Kit, Cleaning Brush, Lapping Tool, Tweezers
Catalog No. 5000580696 Shop All TA Instruments Products
Encompass_Preferred
Waters TA Instruments Discovery DSC 25P Accessory Kit includes 2 silver cell lids, indium calibration material (0.5g), one box each of standard aluminum pans and lids, MDSC sapphire calibration kit, DSC cleaning brush, a cell lapping tool, and stainless steel tweezers.
Catalog No. 50-005-80696 Supplier TA Instruments Supplier No. 973721.901
Waters TA Instruments Discovery DSC 25P Accessory Kit includes 2 silver cell lids, indium calibration material (0.5g), one box each of standard aluminum pans and lids, MDSC sapphire calibration kit, DSC cleaning brush, a cell lapping tool, and stainless steel tweezers.