Learn More
MilliporeSigma™ Supelco™ Discovery™ DSC-NH2 SPE 96-Well Plate, Bed Wt. 100 mg/well
Catalog No. 111008916 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Bed Weight:
100 mg/Well
Catalog No. Bed Weight
11-100-8916 100 mg/Well
1 options
Catalog No. 11-100-8916 Supplier MilliporeSigma™ Supelco™ Supplier No. 575615U
Add to Cart
Add to Cart
Solid phase extraction (SPE) is a technique commonly used in laboratories to isolate analytes of interest from complex matrices.
Retention Mechanism: Normal phase or Anion-exchange. Sample Matrix Compatibility: Organic or aqueous solutions. Polymerically bonded, aminopropyl phase that is very polar in nature (hydrogen bonding) allowing for both normal phase and ion exchange applications. A weak anion exchanger with a pKa of 9.8. At pH 7.8 or below, the functional groups are positively charged. Ion exchange capacity is >= 0.43 meq/g.. Allows the rapid release of very strong anions such as sulfonic acids that may be retained irreversibly by strong anion exchangers. Can be used in some reversed-phase applications (due to ethyl spacer); however, it is predominately used as an ion-exchange or normal phase sorbent due to its polar nature.
TRUSTED_SUSTAINABILITY
Specifications
Column Capacity (Metric) 0.43 meq/g by wetted bed volume
Column Phase DSC-NHs
Column Type SPE
Bed Weight 100 mg/Well
For Use With (Application) Food and Beverages
Product Line Discovery
Type 96-well Plate
Volume (Metric) 2 mL
Endcapped Yes
Material Polypropylene
Pore Size 70 Å pore size
Quantity 1/Pk.
Surface Area (Metric) 480 m 2 /g
Show More Show Less
Discovery is a registered trademark of Sigma-Aldrich Co. LLC