Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Supelco™ Discovery™ DSC-SAX SPE Tube, Bed Wt. 50 mg

Catalog No. 111009379 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Bed Weight:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Bed Weight
11-100-9379 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-100-9379 Supplier MilliporeSigma™ Supelco™ Supplier No. 52661U
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Solid phase extraction (SPE) is a technique commonly used in laboratories to isolate analytes of interest from complex matrices.

Retention Mechanism: Anion-exchangeSample Matrix Compatibility: Organic or aqueous solutions. A polymerically bonded quarternary amine that remains positively charged at all pH levels. Counter ion is Cl-. Ion exchange capacity is >= 0.14 meq/g. Commonly used when extracting weaker anions (e.g., carboxylic acids) that may not bind strongly enough to weaker anion-exchangers. Selectivity can be modified by changing the counter ion with the appropriate buffer during conditioning
TRUSTED_SUSTAINABILITY

Specifications

Column Capacity (Metric) 0.14 meq/g total capacity
Column Phase DSC-SAX
Column Type SPE
Bed Weight 50 mg
For Use With (Application) Food and Beverages
Product Line Discovery
Type Solid Phase Extraction Column
Volume (Metric) 1 mL
Material Polypropylene
Pore Size 70 Å
Quantity 108/Pk.
Surface Area (Metric) 480 m2/g
Show More Show Less

Discovery is a registered trademark of Sigma-Aldrich Co. LLC

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.