Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Supelco™ Discovery™ DSC-SCX SPE Bulk Packing

Catalog No. 111016695 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-6695 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-6695 Supplier MilliporeSigma™ Supelco™ Supplier No. 57221U
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Silica support allows for use with very organic solvents (no shrinking/swelling). Excellent capacity (0.8 meq/g) for cleaning up solution phase combinatorial chemistry reactions (removing target molecules from reaction by-products and excess reagents).

Retention Mechanism: Cation exchange. Sample Matrix Compatibility: Organic or aqueous solutions. A polymerically bonded, benzene sulfonic acid functional group, pKa (< 1.0). Counter ion is H+. Silica support allows for use with very organic solvents (no shrinking/swelling). Excellent capacity (0.8 meq/g) for cleaning up solution phase combinatorial chemistry reactions (removing target molecules from reaction by-products and excess reagents). The presence of the benzene ring offers some mixed-mode capabilities (hydrophobic interactions) that should be considered when extracting cations from aqueous matrices
TRUSTED_SUSTAINABILITY

Specifications

Quantity 100 g
Product Line Discovery

Discovery is a registered trademark of Sigma-Aldrich Co. LLC

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.