Learn More
MilliporeSigma™ Supelco™ Discovery™ DSC-Si SPE 96-Well Plate, Bed Wt. 50 mg/well
Catalog No. 111008949 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Bed Weight:
50 mg/Well
Catalog No. Bed Weight
11-100-8949 50 mg/Well
1 options
Catalog No. 11-100-8949 Supplier MilliporeSigma™ Supelco™ Supplier No. 575608U
Add to Cart
Add to Cart
Solid phase extraction (SPE) is a technique commonly used in laboratories to isolate analytes of interest from complex matrices.
Retention Mechanism: Normal phase. Sample Matrix Compatibility: Organic solvents, oils, and lipids. Unbonded acid washed silica sorbent ideal for normal phase SPE and other modified flash techniques. Often used to separate or remove structurally similar molecules through successive elutions with increasingly polar solutions. The most polar normal phase sorbent available. Excellent capacity for purifying solution phase combinatorial chemistry reactions when removing target molecules from reaction by-products and excess reagents. Available in Büchner Funnel configurations for easy scalability.
TRUSTED_SUSTAINABILITY
Specifications
Column Phase DSC-Si
Column Type SPE
Bed Weight 50 mg/Well
For Use With (Application) Food and Beverages
Product Line Discovery
Type 96-well Plate
Material Polypropylene
Pore Size 70 Å pore size
Quantity 1/Pk.
Surface Area (Metric) 480 m 2 /g
Discovery is a registered trademark of Sigma-Aldrich Co. LLC