Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Supelco™ Discovery™ DSC-WCX SPE Tube, Bed Wt. 100 mg

Catalog No. 111009260 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Bed Weight:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Bed Weight
11-100-9260 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-100-9260 Supplier MilliporeSigma™ Supelco™ Supplier No. 52739U
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Solid phase extraction (SPE) is a technique commonly used in laboratories to isolate analytes of interest from complex matrices.

Retention Mechanism: Cation exchangeSample Matrix Compatibility: Organic or aqueous solutions. A polymerically bonded, ethylenediamine triacetic acid phase with a pKa of 4.8. Counter ion is Na+. Ion exchange capacity is >= 0.15 meq/g. Carries a negative charge at pH 6.8 or above. A pH of 2.8 or below neutralizes this phase for easier elution of strong cationic analytes that are neutralized only at extreme basic conditions. Typically used when dealing with very strong cationic (high pKa) compounds that may be irreversibly retained on strong cation exchangers
TRUSTED_SUSTAINABILITY

Specifications

Column Capacity (Metric) 0.15 meq/g total capacity
Column Phase DSC-WCX
Column Type SPE
Bed Weight 100 mg
For Use With (Application) Food and Beverages
Product Line Discovery
Type Solid Phase Extraction Column
Volume (Metric) 1 mL
Material Polypropylene
Pore Size 70 Å
Quantity 108/Pk.
Surface Area (Metric) 480 m2/g
Show More Show Less

Discovery is a registered trademark of Sigma-Aldrich Co. LLC

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.