Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Discovery Education™ All About Astronomy
SCI_ED

Catalog No. S90456DVD
Change view
Click to view available options
Grade:
3 to 5
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S90456DVD 3 to 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S90456DVD Supplier Discovery Education™ Supplier No. DC880401
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Investigate ways to protect our planet

Examine the technology that has allowed us to explore our solar system and what it's taught us.

15 feature segments, spread across four titles:

  • Protecting Our Planet
  • Liftoff Into Space
  • The Story of the Solar System
  • Exploring Stars

Order Info

Grade 3 to 6. Running time: 2 hours.

Specifications

Grade 3 to 5
Copyright Year 2005
Run Time 120 min.
Name All About Astronomy 4-pack DVD
Format DVD
Includes Marc Record
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.