Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Discovery Education™ Biology: The Science of Life: The Living Cell
SCI_ED

Catalog No. S98140
Change view
Click to view available options
Grade:
6 to 12
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S98140 6 to 12
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S98140 Supplier Discovery Education™ Supplier No. DC010233

missing translation for 'validMainframeMsg'

Help students explore the structure and function of the living cell in both multicellular and unicellular organisms.

Explains that cells are the simplest structures that can carry out all of the activities characteristic of life. Run time: 16 min. Grade Level: 6–12

  • Help student learn about the composition and functions of cells
  • All living things, from simple pond dwelling creatures, like these, to complex animals and plants are made from the same building blocks called cells
  • Cells can reproduce, grow and develop, respond to their surroundings, and so on
  • Cells are like miniature factories which use raw materials and energy to create their amazing product which is none other than life itself
  • Large organisms are multicellular and are made from many different cells
  • The cells of multicellular organisms have become specialized to perform all sorts of tasks such as those carried out by muscles and blood
  • Many of the smallest organisms, such as the Protist paramecia, are unicellular and possess just one cell

Specifications

Grade 6 to 12
Format DVD
Includes Marc Record
Name Biology: the Science of Life: the Living Cell DVD
Run Time 16 min.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.