Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Discovery Education™ Galapagos: Beyond Darwin
SCI_ED

Catalog No. S87753
Change view
Click to view available options
Grade:
6 to 12
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S87753 6 to 12
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S87753 Supplier Discovery Education™ Supplier No. DC716068
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Groundbreaking research project picks up where Darwin left off

Deep beneath the ocean's surface live creatures Darwin could only have imagined in the 1800s.

Order Info

Grades 6 to 12. Running time: 52 mins.

Specifications

Grade 6 to 12
Copyright Year 1997
Name Galapagos: Beyond Darwin VHS
Includes Marc Record, Teacher's Guide
Run Time 52 min.
Format VHS
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.