Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Discovery Education™ Great Inventions with Bill Nye: Communication
SCI_ED

Catalog No. s95447
Change view
Click to view available options
Grade:
6 to 12
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S95447 6 to 12
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S95447 Supplier Discovery Education™ Supplier No. DC903617
Only null left
Add to Cart
Add to Cart

Retrace the innovations that allow humans to contact almost anyone, anywhere, at any time.

Bill Nye outlines a number of technological advancements that have enabled humans to use both written and spoken language to communicate faster, farther, and with more people than ever before.

  • Grades 6 to 12
  • 56 mins.

Specifications

Grade 6 to 12
Copyright Year 2007
Name Greatest Inventions with Bill Nye: Communication VHS
Run Time 56 min.
Includes Marc Record, Teacher's Guide
Format VHS
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.