Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Discovery Education™ Learning with Animals Five-Pack
SCI_ED

Catalog No. S90455
Change view
Click to view available options
Grade:
3 to 5
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S90455 3 to 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S90455 Supplier Discovery Education™ Supplier No. DC879148
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

Grade K to 2. Running time: 1 hour, 25 mins.

Use children's love of animals as a springboard for teaching everything from letter and number basics to vocabulary and physical fitness

Includes five titles:

  • Animal Alphabet
  • Animals Around Us
  • Animal 1-2-3
  • Animal Colors and Shapes
  • Animals in Action

Specifications

Grade 3 to 5
Copyright Year 2005
Includes Marc Record
Name Learning with Animals 5-pack VHS
Format VHS
Run Time 85 min.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.