Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Discovery Education™ MyPyramid: Simple Steps to Healthy Living DVD
SCI_ED

Catalog No. S98529VHS
Change view
Click to view available options
Grade:
3 to 8
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S98529VHS 3 to 8
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S98529VHS Supplier Discovery Education™ Supplier No. DC023847
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

teacher's Guide

How can every individual achieve the right balance of proper nutrition and exercise?

Learn the importance and use of the USDA MyPyramid food guidelines. Run Time: 15 min. Grades 9 to 12.

  • Explore the six basic food groups
  • See how families make sure they eat enough whole grains, fruits, and vegetables

Specifications

Grade 3 to 8
Copyright Year 1990
Name Mypyramid: Simple Steps to Healthy Living DVD
Format DVD
Includes Marc Record
Run Time 15 min.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.