Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Discovery Education™ MythBusters: Forces
SCI_ED

Catalog No. S95466DVD
Change view
Click to view available options
Grade:
6 to 12
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S95466DVD 6 to 12
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S95466DVD Supplier Discovery Education™ Supplier No. DC902866
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Is it possible for a human being to fall from a jet airliner in flight and live?

Could you survive being hit on the head by a penny dropped from the top of a skyscraper? Adam Savage and Jamie Hyneman explore the gravity of several myths.

  • Grades 6-12
  • 24 mins.

Specifications

Grade 6 to 12
Run Time 24 min.
Format DVD
Copyright Year 2007
Includes Marc Record, Teacher's Guide
Name Mythbusters: Forces DVD
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.