Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Discovery Education™ Planet Earth: The Future: Living Together
SCI_ED

Catalog No. S98075
Change view
Click to view available options
Grade:
6 to 12
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S98075 6 to 12
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S98075 Supplier Discovery Education™ Supplier No. DC020271
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Study the conservation movement in today's world and the implications of sustainable development on overall growth of society.

The 2008 AEP Distinguished Achievement award-winning program shows Planet Earth in never-before-seen biological and ecological perspectives and discusses how to maintain the balance between conservation and economic expansion. Run time: 46 min. Grade Level: 9dash (en)12.

  • Journey through the rain forests of Borneo and the peaks of the Himalayas to uncover the beauty and mystery of life on our planet
  • Watch never-before-seen animal behaviors and startling views of locations and ecosystems
  • Understand all about conservation in the wake of destruction and ecotourism and responsible farming
  • Become aware of the big game hunting in Africa and the destruction of rain forests in Borneo and other ethical dilemmas affecting the environment
  • Watch naturalists, economists and scientists discuss about preserving the environment while maintaining economic expansion
  • Learn about the struggle and the progress made in preserving Earth's most vital ecosystems

Specifications

Grade 6 to 12
Run Time 50 min.
Format VHS
Includes Marc Record
Name Planet Earth: The Future: Living Together VHS
Copyright Year 2007
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.