Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Discovery Education™ Prehistoric Earth VHS/DVD
SCI_ED

Catalog No. S95991DVD
Change view
Click to view available options
Grade:
3 to 5
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S95991DVD 3 to 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S95991DVD Supplier Discovery Education™ Supplier No. DC778613
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ever wonder how geologists know what happened on Earth billions of years ago?

Video covers the nature and divisions of geologic time, a variety of prehistoric life-forms, and how fossils reveal clues to Earth's distant past. Grades 3-6. Running time: 25 minutes.

Geologic Time

  • Explains the system used by scientists to describe and organize prehistoric time periods

Prehistoric Life

  • Discusses what modern scientists study to uncover facts about the past

Events in Earth's Past

  • Chronicles events in the four major time periods (eras) within the geologic time scale: Precambrian, Paleozoic, Mesozoic, and Cenozoic

Dinosaurs and Fossils

  • Points out that each time period is marked by a significant change in the number or type of fossils that are found
  • Explores what fossils reveal about Earth's history

Specifications

Grade 3 to 5
Copyright Year 2005
Name Prehistoric Earth DVD
Format DVD
Run Time 25 min.
Includes Marc Record, Teacher's Guide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.