Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Discovery Education™ Rocks and Minerals Learning Pack
SCI_ED

Catalog No. S68562
Change view
Click to view available options
Format:
CD-ROM
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Format
S68562 CD-ROM
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S68562 Supplier Discovery Education™ Supplier No. DC770123
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

  • Windows™ and Mac™ compatible, elementary age-appropriate CD-ROM with interactive games, activities, and multimedia tools
  • 30-pack classroom set of Discovery Wraps student worksheets
  • Comprehensive Teacher's Guide book with lesson plans and teaching strategies
  • Free online lesson support

Learn the differences between igneous, sedimentary, and metamorphic rocks; study lava; and more.

Video appropriate for elementary school students; three segments with a total running time of 26 minutes.

Order Info

Grades 3-6

Specifications

Grade 3 to 6
Compatibility Windows™, Mac™
Category Earth Science
Format CD-ROM
For Use With (Application) Learn the differences between igneous, sedimentary, and metamorphic rocks, study lava, and more
Includes Elementary age appropriate CD-ROM Interactive games, activities, and multimedia tools, 30-pack classroom set of Discovery Wraps student worksheets, comprehensive teacher's guide book with lesson plans and teaching strategies, free online lesson support
Product Type Rocks and Minerals Learning Pack
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.