Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Discovery Education™ Stuff Happens with Bill Nye - Breakfast DVD
SCI_ED

Catalog No. S98528DVD
Change view
Click to view available options
Grade:
6 to 12
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S98528DVD 6 to 12
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S98528DVD Supplier Discovery Education™ Supplier No. DC024052
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Learn how everyday stuff of our lives and the far-reaching effects our actions have on the planet.

It offers alternatives to these global problems through small changes each person can make to create a huge difference in the health of the planet. Running time: 22 min., Grade 6 to 12.

  • Explains how technology can help or hurt the environment, and actions we can implement to improve our use of resources
  • Bill Nye points out how choices in breakfast foods can be some of the most important for the environment
  • From production to shipping, the program travels around the world to examine how food can place a heavy burden on the earth's natural resources
  • Teachers can utilize the keen delivery of Bill Nye to help students understand human interaction with the environment

Specifications

Grade 6 to 12
Copyright Year 2008
Name Stuff Happens with Bill Nye-Breakfast DVD
Format DVD
Run Time 22 min.
Includes Marc Record
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.