Learn More
TA Instruments Discovery Finned Air Cooling System (FACS), Quench or Programmed Cooling of DSC Cell from 725°C to Ambient Temperatures, Uses Compressed Air or Nitrogen, Compatible with Discovery DSC Series
Catalog No. 5000580710 Shop All Waters TA Instruments Products
Encompass_Preferred
Waters TA Instruments Discovery Finned Air Cooling System (FACS) provides quench or programmed cooling of Discovery Series DSC cells from 725°C to ambient using customer-supplied compressed gas (air or nitrogen) as the cooling medium.
Catalog No. 50-005-80710 Supplier TA Instruments Supplier No. 972503.901
Waters TA Instruments Discovery Finned Air Cooling System (FACS) provides quench or programmed cooling of Discovery Series DSC cells from 725°C to ambient using customer-supplied compressed gas (air or nitrogen) as the cooling medium.