Learn More
TA Instruments Discovery LN Pump, Liquid Nitrogen Cooling, Lowest Temperature -180°C, Cooling Rate up to 140°C/min, Auto-Fill Dewar, Software Controlled, Compatible with Discovery DSCs and X3 DSC
Catalog No. 5000580757 Shop All Waters TA Instruments Products
Encompass_Preferred
Waters TA Instruments Discovery Liquid Nitrogen (LN) Pump for the Discovery DSC series provides the highest performance and greatest flexibility in cooling. It facilitates the lowest operational temperature (-180°C), greatest cooling rate capacity (to 140°C/min), fastest sub-ambient equilibration times, and an upper-temperature limit of 550 °C. Operating at ambient pressure, it uses liquid nitrogen efficiently, reducing operating costs. It features a 50-liter Dewar with self-filling capability and software-controlled auto-fill during experiments. Compatible With Discovery DSCs and the X3 DSC.
Catalog No. 50-005-80757 Supplier TA Instruments Supplier No. 973900.901
Waters TA Instruments Discovery Liquid Nitrogen (LN) Pump for the Discovery DSC series provides the highest performance and greatest flexibility in cooling. It facilitates the lowest operational temperature (-180°C), greatest cooling rate capacity (to 140°C/min), fastest sub-ambient equilibration times, and an upper-temperature limit of 550 °C. Operating at ambient pressure, it uses liquid nitrogen efficiently, reducing operating costs. It features a 50-liter Dewar with self-filling capability and software-controlled auto-fill during experiments. Compatible With Discovery DSCs and the X3 DSC.