Learn More
TA Instruments Discovery PCA Trigger Adapter, Communication and Mounting Kit, Converts USB to RS232 for PCA Operation with First Generation Discovery DSC
Catalog No. 5000580637 Shop All Waters TA Instruments Products
Encompass_Preferred
Waters TA Instruments Discovery PCA Trigger Adapter enables communication between first generation Discovery DSC instruments and the PCA by converting USB signals to RS232. The kit includes both the communication adapter and Discovery PCA Adaptor Ring (P/N: 935050.001), required for correct mounting of the PCA Light Guide Adaptor Base. This accessory ensures compatibility and functionality of PCA accessories with earlier Discovery DSC models. By maintaining reliable signal communication and mechanical alignment, the adapter provides laboratories with a cost-effective solution for integrating modern PCA functionality with legacy instruments.
Catalog No. 50-005-80637 Supplier TA Instruments Supplier No. 972734.901
Waters TA Instruments Discovery PCA Trigger Adapter enables communication between first generation Discovery DSC instruments and the PCA by converting USB signals to RS232. The kit includes both the communication adapter and Discovery PCA Adaptor Ring (P/N: 935050.001), required for correct mounting of the PCA Light Guide Adaptor Base. This accessory ensures compatibility and functionality of PCA accessories with earlier Discovery DSC models. By maintaining reliable signal communication and mechanical alignment, the adapter provides laboratories with a cost-effective solution for integrating modern PCA functionality with legacy instruments.