Learn More
TA Instruments Discovery TGA 55 EGA, Vertical Thermo-Balance, Dual Range Microbalance, Gas Purge System, EGA Furnace, Touch Screen, Gas Delivery Manifold, TRIOS Software, Sample Kit, 5-Year Furnace Warranty, Windows 7 +
Catalog No. 5000580712 Shop All Waters TA Instruments Products
Encompass_Preferred
Waters TA Instruments Discovery TGA 55-EGA includes a sensitive vertical thermo-balance, dual range microbalance, horizontal purge system, EGA furnace for FTIR, MS, GC analysis, touch screen, gas delivery manifold, TRIOS software, accessory kit, and 5-year furnace warranty. Compatible with Windows 7+, DSL router required.
Catalog No. 50-005-80712 Supplier TA Instruments Supplier No. 956800.901
Waters TA Instruments Discovery TGA 55-EGA includes a sensitive vertical thermo-balance, dual range microbalance, horizontal purge system, EGA furnace for FTIR, MS, GC analysis, touch screen, gas delivery manifold, TRIOS software, accessory kit, and 5-year furnace warranty. Compatible with Windows 7+, DSL router required.