Learn More
TA Instruments Discovery TGA 55, Vertical Thermo-Balance, Dual Range Microbalance, Gas Purge System, Touch Screen, Gas Delivery Manifold, TRIOS Software, Sample Kit, 5-Year Furnace Warranty, Windows 7 +
Catalog No. 5000580901 Shop All Waters TA Instruments Products
Encompass_Preferred
Waters TA Instruments Discovery TGA 55 features a sensitive vertical thermo-balance with dual range microbalance 0-200 mg/0-1000 mg, horizontal gas purge system, touch screen, gas delivery manifold, TRIOS software, sample kit, calibration materials, tools, and 5-year furnace warranty. Compatible with Windows 7 and greater, DSL router required.
Catalog No. 50-005-80901 Supplier TA Instruments Supplier No. 956450.901
Waters TA Instruments Discovery TGA 55 features a sensitive vertical thermo-balance with dual range microbalance 0-200 mg/0-1000 mg, horizontal gas purge system, touch screen, gas delivery manifold, TRIOS software, sample kit, calibration materials, tools, and 5-year furnace warranty. Compatible with Windows 7 and greater, DSL router required.