Learn More
TA Instruments Discovery TGA 550 55 Mass Spectrometer Interface Kit Without Library, TGA Clamping Bracket and Fittings Included, Compatible with Discovery MS and Pfeiffer Vacuum Thermostar MS, Wiley NIST Library Optional
Catalog No. 5000580820 Shop All Waters TA Instruments Products
Encompass_Preferred
Waters TA Instruments Discovery TGA 550/55 Mass Spectrometer Interface Kit without library is designed for use with an EGA furnace. The kit includes a TGA clamping bracket and fittings and is compatible with the Discovery MS and the Pfeiffer Vacuum Thermostar MS. The Wiley/NIST library of mass spectral data is available separately as an optional item.
Catalog No. 50-005-80820 Supplier TA Instruments Supplier No. 271001.002
Waters TA Instruments Discovery TGA 550/55 Mass Spectrometer Interface Kit without library is designed for use with an EGA furnace. The kit includes a TGA clamping bracket and fittings and is compatible with the Discovery MS and the Pfeiffer Vacuum Thermostar MS. The Wiley/NIST library of mass spectral data is available separately as an optional item.