Learn More
Roche Diagnostics DISCOVERY Wash (10X), 2 L
Supplier:  Roche Diagnostics 07311079001
Encompass
The ROCHE DIAGNOSTIC DISCOVERY Wash (10X) (RUO) is a concentrated wash solution tailored for research applications, featuring a 10X concentration that makes it essential for various experimental designs. This buffer is specifically crafted to enhance the performance of automated immunohistochemistry (IHC), in situ hybridization (ISH), and multiplexing procedures, thereby ensuring more reliable washing steps. This product is exclusively for research use and is not intended for diagnostic purposes, catering to the needs of scientists and researchers seeking dependable solutions for their workflows.
  • 10X concentrated wash solution.
  • Designed for research applications only.
  • Compatible with automated IHC, ISH, and multiplexing.
  • Ensures reliable washing performance.
  • Suitable for use with DISCOVERY ULTRA and XT systems.
  • Volume: 2 L.
Catalog No. NC2072541