Learn More
STEMCELL Technologies DisCoZyme 2, ACF, Size: 50 mg
SDP
Catalog No. 501974408
Encompass
Collagenase/dispase blend with high collagenase and high caseinase activity
Catalog No. 50-197-4408 Supplier STEMCELL Technologies Supplier No. 07450
Collagenase/dispase blend with high collagenase and high caseinase activity