Learn More
Medchemexpress LLC Diselenide, bis[3-(trifluoromethyl)phenyl] | 53973-75-4 | 100.0% | 448.12 g·mol⁻¹ | C14H8F6Se2 | 10 MG

Supplier: Medchemexpress LLC HY4730810MG
1,2-Bis(3-(trifluoromethyl)phenyl)diselane is a diselenide research reagent provided in small-milligram quantities and as a DMSO solution. It appears as a light yellow liquid with high reported purity and requires storage under inert atmosphere at low temperature for stability.
- High reported purity (≈99.97%).
- Light yellow liquid physical form.
- Molecular formula C14H8F6Se2; molecular weight 448.12 g·mol⁻¹.
- Available in small pack sizes and as a 10 mM solution in DMSO.
- Store at -20°C under nitrogen; in solution store at -80°C for long-term.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.