Learn More
Cellvis DISH 35MM 10/SLV 100CS
Catalog No. NC0699576
Encompass
35mm glass bottom dish, dish size 35mm, well size 20mm, #1 cover glass(0.13mm-0.16mm).
Catalog No. NC0699576 Supplier Cellvis Supplier No. D35201N
35mm glass bottom dish, dish size 35mm, well size 20mm, #1 cover glass(0.13mm-0.16mm).