Learn More
Cellvis DISH,35MM W/1.5 GLASS BOTTOM
Catalog No. NC0409658
Encompass
35mm glass bottom dish, dish size 35 mm, well size 20mm, #1.5 cover glass(0.16-0.19mm). 100 per case.
Catalog No. NC0409658 Supplier Cellvis Supplier No. D35201.5N
35mm glass bottom dish, dish size 35 mm, well size 20mm, #1.5 cover glass(0.16-0.19mm). 100 per case.