Learn More
Hach Company Dish, Aluminum, Disposable
Supplier:  Hach Company 2164000
Encompass
Aluminum dish for evaporating and general utility use. Crimped sides for stability. Sturdy, with a flat bottom and slight taper for easy stacking. Dished include tabs for easy handling. Temperature range 0 to 426°C (800°F). Melting point 676°C (1250°F). Dish height approx. 3/4".
Catalog No. 50-199-5486